Skip to Content
Merck
All Photos(1)

Key Documents

FCMAB187F

Sigma-Aldrich

Milli-Mark® Anti-CD23-FITC Antibody, clone MHM6

clone MHM6, Milli-Mark®, from mouse

Synonym(s):

Low affinity immunoglobulin epsilon Fc receptor, Lymphocyte IgE receptor, Fc-epsilon-RII, BLAST-2, Immunoglobulin E-binding factor, C-type lectin domain family 4 member J

Sign Into View Organizational & Contract Pricing

Select a Size

100 TESTS
€403.00

€403.00


Estimated to ship on23 April 2025


Request a Bulk Order

Select a Size

Change View
100 TESTS
€403.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.44

€403.00


Estimated to ship on23 April 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

FITC conjugate

antibody form

purified antibody

antibody product type

primary antibodies

clone

MHM6, monoclonal

species reactivity

human

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FCER2(2208)

General description

CD23 is a 45 kDa type II membrane glycoprotein belonging to the C-type lectin family, and is identical to the low affinity IgE receptor (FceRII) (Kikutani, 1987; Aubry, 1987). CD23 is primarily expressed on B cells and monocytes including a strong expression on EBV-transformed B lymphoblasts (Sarfati, 1997; Sarfati, 1995). CD23 is also present on a large variety of other cells such as T cells, eosinophils, platelets, Langerhans cells, and a subset of thymic epithelial cells (Sarfati, 1995). However, CD23 is absent in a large number of normal non-lymphoid human tissues, such as, central nervous system, endocrine glands, gastrointestinal, respiratory, urinary, and female/male reproductive system, skin, bone, cartilage, connective tissue, muscles, peripheral nerves, vessels, mesothelium, synovium, placenta, and umbilical cord (Pallesen, 1987). Anti-CD23, MHM6, was included in the Second, Third, Fourth, and Fifth International Workshop and Conference on Human Leukocyte Differentiation Antigens, and studies by a number of laboratories confirmed its reactivity with CD23 (Sarfati, 1995).

Specificity

Antibody recognizes Human CD23 antigen.

Immunogen

EBV-transformed B-lymphoblastoid cells

Application

Detect CD23 using this Milli-Mark Anti-CD23-FITC Antibody, clone MHM6 validated for use in FC.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Quality

Evaluated by flow cytometry using human lymphocytes.

Target description

45 kDa (Kikutani, 1987; Aubry, 1987)

Physical form

Protein A purified
Purified Mouse monoclonal IgG1 conjugated to FITC in 0.05M Tris-HCl buffer with 1% BSA, Sodium Azide at 0.097% at pH 7.2.

Storage and Stability

Maintain refrigerated at 2-8°C protected from light in undiluted aliquots for up to 6 months from date of receipt.

Analysis Note

Control
human lymphocytes

Legal Information

MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service