Skip to Content
Merck
All Photos(4)

Documents

ABS31

Sigma-Aldrich

Anti-2A Peptide Antibody

serum, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity (predicted by homology)

all

technique(s)

western blot: suitable

shipped in

wet ice

target post-translational modification

unmodified

General description

2A peptides and 2A-like peptide sequences (also known as CHYSEL, or cis-acting hydrolase elements) are a superior alternative to internal ribosomal entry sites (IRES) for coordinating the expression of multiple gene products from a single recombinant construct. 2A peptides allow multiple proteins to be encoded as polyproteins, which then dissociate into component proteins upon translation. The 2A sequence impairs normal peptide bond formation via a mechanism called ribosomal skipping, resulting in effective, non-enzymatic generation of distinct peptide products from a single multicistronic construct. The use of 2A sequences allows the stoichiometric production of up to four proteins from a single vector, making it a powerful tool for the equimolar expression of multiple cistrons.

Specificity

Recognizes the 2A sequence, derived from Foot and Mouth picornavirus (VKQTLNFDLLKLAGDVESNPG*P), where * represents the 2A cleavage site. The epitope is believed to be within the “GDVESNPG” region since the antibody also recognizes the unrelated 2A regions from Thosea asigna virus and Equine rhinitis A virus.

Immunogen

Epitope: The epitope is believed to be within the “GDVESNPG” region.
The immunogen used was recombinant 2A from foot and mouth virus1.

Application

Anti-2A Peptide Antibody is an antibody against 2A Peptide for use in WB.
Research Category
Epitope Tags & General Use
Research Sub Category
Epitope Tags

Quality

Western Blot Analysis:
A multicistronic vector was synthesized consisting of several CD3 proteins linked with 2A peptide sequences from foot and mouth disease, Thosea asigna, and Equine rhinitis A viruses, respectively (CD3δ-F2A-CD3ε-T2A-CD3γ-E2A-CD3ζ)1. The vector was transduced in 293 cells. A 1:1000 dilution of this antibody detected the 2A sequence in all three CD3-2A proteins (CD3 epsilon, gamma, delta) generated from a single 2A peptide-linked retroviral vector.

Target description

varies

Linkage

Replaces: 09-085

Physical form

Serum with 0.05% NaN3.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
2A-containing recombinant proteins

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evamaria Gruchattka et al.
PloS one, 10(12), e0144981-e0144981 (2015-12-25)
Engineering of the central carbon metabolism of Saccharomyces cerevisiae to redirect metabolic flux towards cytosolic acetyl-CoA has become a central topic in yeast biotechnology. A cell factory with increased flux into acetyl-CoA can be used for heterologous production of terpenoids
Xavier Meckler et al.
The Journal of biological chemistry, 291(24), 12821-12837 (2016-04-10)
γ-Secretase complexes achieve the production of amyloid peptides playing a key role in Alzheimer disease. These proteases have many substrates involved in important physiological functions. They are composed of two constant subunits, nicastrin and PEN2, and two variable ones, presenilin
Andrew Hofacre et al.
Human gene therapy, 29(4), 437-451 (2017-12-09)
Toca 511, a retroviral replicating vector (RRV), uses an internal ribosomal entry site (IRES) to express an optimized yeast cytosine deaminase (yCD2), which converts 5-fluorocytosine to 5-fluorouracil. This configuration is genetically stable in both preclinical mouse models and human clinical
Processing of the VP1/2A junction is not necessary for production of foot-and-mouth disease virus empty capsids and infectious viruses: characterization of "self-tagged" particles.
Gullberg, M; Polacek, C; B?tner, A; Belsham, GJ
Journal of virology null
Maria Gullberg et al.
PloS one, 11(6), e0157435-e0157435 (2016-06-15)
Foot-and-mouth disease (FMD) remains one of the most economically important infectious diseases of production animals globally. Vaccination can successfully control this disease, however, current vaccines are imperfect. They are made using chemically inactivated FMD virus (FMDV) that is produced in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service