Skip to Content
Merck
All Photos(1)

Key Documents

ABS2183M

Sigma-Aldrich

Anti-Coagulation factor VIIa

from rabbit, purified by affinity chromatography

Synonym(s):

Coagulation factor VII, EC: 3.4.21.21, Proconvertin, Serum prothrombin conversion accelerator, SPCA

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
€131.00
100 μG
€320.00

€131.00


Estimated to ship on14 April 2025


Request a Bulk Order

Select a Size

Change View
25 μG
€131.00
100 μG
€320.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€131.00


Estimated to ship on14 April 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

primate (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... F7(2155)

General description

Coagulation factor VII (UniProt: P08709; also known as EC: 3.4.21.21, Proconvertin, Serum prothrombin conversion accelerator, SPCA) is encoded by the F7 gene (Gene ID: 2155) in human. Coagulation factor VII is a member of the peptidase S1 family that initiates the extrinsic pathway of blood coagulation. It is serine protease that is synthesized with a signal peptide (aa 1-20) and a propeptide (aa 21-60) and circulates in the blood in a zymogen form. The mature form is a heterodimer of a light and a heavy chain. It is a vitamin K-dependent protease that binds to calcium following enzymatic carboxylation of some glutamate residues. Factor VII is converted to Factor VIIa by the action of Factor Xa, Factor XIIa, Factor IXa, or thrombin by minor proteolysis. In the presence of tissue factor and calcium ions, Factor VIIa converts factor X to factor Xa by cleavage of Arg-|-Ile bond. Factor VIIa also converts factor IX to factor IXa in the presence of tissue factor and calcium. Factor VII deficiency resulting from mutations in F7 gene can lead to hemorrhagic complications.

Specificity

This rabbit polyclonal antibody detects coagulation factor VIIa in human.

Immunogen

Full-length human recombinant Coagulation factor VIIa.

Application

Anti-Coagulation factor VIIa Antibody, Cat. No. ABS2183, is a rabbit polyclonal antibody that detects Coagulation factor VIIa and has been tested for use in Immunocytochemistry, Immunofluorescence, and Western Blotting.
Immunofluorescence Analysis: A representative lot detected Coagulation factor VIIa in endothelial cell cultures incubated with rFVIIa (6 mg/mL) for 2 h at 37˚C.

Immunocytochemistry Analysis: A representative lot detected Coagulation factor VIIa in umbilical cord veins.

Quality

Evaluated by Western Blotting in human placental tissue lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected Coagulation factor VIIa in 10 µg of human placental tissue lysate.

Target description

~57 kDa observed; 51.59 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service