Skip to Content
Merck
All Photos(3)

Key Documents

AB9986

Sigma-Aldrich

Anti-phospho-BRCA2 (Ser3291) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

BRCA1/BRCA2-containing complex, subunit 2, Fanconi anemia group D1 protein 3, Fanconi anemia, complementation group D1, breast and ovarian cancer susceptibility gene, early onset, breast cancer 2 tumor suppressor, breast cancer 2, early onset, breast can

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€421.00

€421.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€421.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€421.00


Estimated to ship on23 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rhesus macaque, horse, human

species reactivity (predicted by homology)

rat (based on 100% sequence homology), mouse (based on 100% sequence homology), hamster (based on 100% sequence homology), cat (based on 100% sequence homology), equine (based on 100% sequence homology), canine (based on 100% sequence homology), chimpanzee (based on 100% sequence homology), rhesus monkey (based on 100% sequence homology)

technique(s)

dot blot: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer3291)

Gene Information

human ... BRCA2(675)

General description

BRCA2 (Breast Cancer Type 2 susceptibility protein) is a tumor suppressor protein encoded by the BRCA2 gene.
The protein encoded by this gene plays a direct role in error-free double-strand break repair and homologous recombination. BRCA2 shares similarities with the tumor suppressor gene BRCA1. Inherited mutations in BRCA2 as well as BRCA1 are associated with an increased risk of developing breast or ovarian cancer. Although the structures of the BRCA1 and BRCA2 genes are very different, some functions are similar. The proteins made by both genes are essential for repairing damaged DNA and interact with the protein produced by the RAD51 gene. Together these three proteins play a role in maintaining the stability of the human genome and prevent dangerous gene rearrangements that can lead to the development of cancer.

Specificity

This antibody recognizes human BRCA2 phosphorylated at and around the internal domain surrounding Ser3291.

Immunogen

Epitope: Internal Domain surrounding Ser3291
KLH-conjugated linear peptide corresponding to human BRCA2 phosphorylated at and around the internal domain surrounding Ser3291.

Application

Anti-phospho-BRCA2 (Ser3291) Antibody is a rabbit polyclonal antibody for detection of phospho-BRCA2 (Ser3291), also known as BRCA1/BRCA2-containing complex subunit 2 & Fanconi anemia group D1 protein 3, has been validated in ICC, IHC, DB.
Immunocytochemistry Analysis: 1:500 dilution from a previous lot detected BRCA2 in MCF-7 cells.

Immunohistochemistry Analysis: 1:100 dilution from a previous lot detected BRCA2 in metastatic ductal carcinoma tissue.
Research Sub Category
Cell Cycle, DNA Replication & Repair

Oncoproteins

Quality

Evaluated by Dot Blot in modified and non modified peptides of phosphorylated and non-phosphorlyated BRCA2.

Dot Blot Analysis: 2 µg/mL of this antibody detected BRCA2 in modified and non modified peptides of phosphorylated and non-phosphorlyated BRCA2.

Target description

~ 390 kDa

Analysis Note

Control
Modified and non modified peptides of phosphorylated and non-phosphorlyated BRCA2

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lifeng Chen et al.
Clinical and translational medicine, 11(3), e341-e341 (2021-03-31)
PARP inhibitors induce DNA lesions, the repair of which are highly dependent on homologous recombination (HR), and preferentially kill HR- deficient cancers. However, cancer cells have developed several mechanisms to transform HR and confer drug resistance to PARP inhibition. Therefore
Fang Zhang et al.
Cell reports, 32(4), 107974-107974 (2020-07-30)
Attenuated DNA repair leads to genomic instability and tumorigenesis. BRCA1/BARD1 are the best-known tumor suppressors that promote homology recombination (HR) and arrest cell cycle. However, it remains ambiguous whether and how their E3 ligase activity regulates HR. Here, we demonstrate
Gro Elise Rødland et al.
Frontiers in oncology, 9, 1301-1301 (2019-12-19)
The CD37 targeting radioimmunoconjugate 177Lu-lilotomab satetraxetan (Betalutin) is currently being evaluated in a clinical phase 2b trial for patients with follicular lymphoma (FL) and in a phase 1 trial for patients with diffuse large B-cell lymphoma (DLBCL). Herein we have
Sissel Hauge et al.
Oncotarget, 8(7), 10966-10979 (2016-12-29)
Recent studies have shown synergistic cytotoxic effects of simultaneous Chk1- and Wee1-inhibition. However, the mechanisms behind this synergy are not known. Here, we present a flow cytometry-based screen for compounds that cause increased DNA damage in S-phase when combined with
Gro Elise Rødland et al.
Cancers, 13(15) (2021-08-08)
Inhibitors of WEE1 and ATR kinases are considered promising for cancer treatment, either as monotherapy or in combination with chemo- or radiotherapy. Here, we addressed whether simultaneous inhibition of WEE1 and ATR might be advantageous. Effects of the WEE1 inhibitor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service