Skip to Content
Merck
All Photos(1)

Documents

AB5372

Sigma-Aldrich

Anti-Trk B Antibody, pain, a.a. 54-67 rTrkB

serum, Chemicon®

Synonym(s):

BDNF/NT-3 Growth Factors Receptor, Tyrosine Kinase Receptor B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... NTRK2(4915)

Specificity

TrkB. The sequence used for the immunogen was chosen to avoid cross reactivity with trkA or trkC.

Immunogen

A synthetic peptide from rat trkB
Epitope: a.a. 54-67 rtrkB

Application

Anti-Trk B Antibody, pain, a.a. 54-67 rTrkB detects level of Trk B & has been published & validated for use in IH & WB.
Immunohistochemistry: 1:2,000-1:3,000. The antibody reacts well with tissue from the rat spinal cord.
Immunoblotting: 1:1,000-1:1,500 on rat membrane preparations. The antibody should be
evaluated for use in Western blot by the end user. Its utility will be dependant on the level of
the protein expressed in the tissues or cells being tested.
Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Neurochemistry & Neurotrophins

Physical form

Rabbit serum. Lyophilized. Contains no preservative. Reconstitute with 50 μL of sterile distilled water. Centrifuge if necessary to remove any residue.

Storage and Stability

Maintain lyophilized material at -20°C to -70°C for up to 6 months after date of receipt. After reconstitution maintain frozen at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zitao Zhang et al.
Molecular medicine reports, 15(3), 1362-1367 (2017-01-19)
Brain‑derived neurotrophic factor (BDNF), a member of the neurotropic family, is expressed in osteoblast‑like cells of a fracture callus, however, its role in fracture healing remains to be fully elucidated. Osteoblasts isolated from Sprague Dawley rats were stimulated by BDNF
Laurens W J Bosman et al.
Brain cell biology, 35(1), 87-101 (2007-10-18)
The receptor tyrosine kinase TrkB and its ligands, brain-derived neurotrophic factor (BDNF) and neurotrophin-4/5 (NT-4/5), are critically important for growth, survival and activity-dependent synaptic strengthening in the central nervous system. These TrkB-mediated actions occur in a highly cell-type specific manner.
Rita Machaalani et al.
Neurotoxicology, 74, 40-46 (2019-05-24)
Cigarette smoke exposure during pregnancy into infancy affects brain growth and development in both short and long term (into adulthood). Using a mouse model of pre- into post- natal cigarette smoke exposure (SE), we aimed to determine the effects on
Ji Min Yu et al.
Methods in molecular biology (Clifton, N.J.), 702, 219-231 (2010-11-18)
While adult stem cells can be induced to transdifferentiate into multiple lineages of cells or tissues, their plasticity and utility for human therapy remains controversial. In this chapter, we describe methods for the transdifferentiation of human adipose tissue-derived stem cells
Amy A Robinson et al.
GeroScience, 40(1), 31-47 (2018-01-23)
Mammalian aging is associated with decline in cognitive functions. Studies searching for a cause of cognitive aging initially focused on neuronal loss but quantitative investigations of rat, monkey, and human brain using stereology demonstrated that in normal aging, unlike in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service