Skip to Content
Merck
All Photos(1)

Documents

8.20219

Sigma-Aldrich

Ethyl 2-bromopropionate

for synthesis

Synonym(s):

Ethyl 2-bromopropionate, 2-Bromopropionic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9BrO2
CAS Number:
Molecular Weight:
181.03
MDL number:
UNSPSC Code:
12352108
EC Index Number:
208-609-5

Assay

≥98.0% (GC)

form

liquid

potency

281 mg/kg LD50, oral (Rat)

bp

156-160 °C/1013 hPa

transition temp

flash point 52 °C

density

1.39 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C5H9BrO2/c1-3-8-5(7)4(2)6/h4H,3H2,1-2H3

InChI key

ARFLASKVLJTEJD-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.388 - 1.393
Identity (IR): passes test

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(C)

51 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service