Skip to Content
Merck
All Photos(1)

Documents

6.18663

1-Propanol

for chromatography

Synonym(s):

1-Propanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H8O
CAS Number:
Molecular Weight:
60.10
MDL number:
UNSPSC Code:
12191601

grade

for chromatography

Quality Level

InChI

1S/C3H8O/c1-2-3-4/h4H,2-3H2,1H3

InChI key

BDERNNFJNOPAEC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Propanol(n-propyl alcohol) is a short-chain alcohol, used as an extraction solvent inhigh-performance liquid chromatography (HPLC) or gas chromatography (GC). Itcan be used as a mobile-phase solvent or as a solvent for sample preparation,dilution, or extraction in chromatographic analyses. It is also a highlyflammable, colorless liquid, used as a solvent for printing inks and as achemical intermediate.

Application

1-Propanol can be used as a mobile phase component in:
  • SDS-mediated chromatographic system to analyze the chromatographic behavior of β-blockers in a reversed-phase liquid chromatographic (RPLC) column.
  • Separation of taurine during the detection of adulteration in an edible bird′s nest.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

71.6 °F - closed cup

Flash Point(C)

22 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

n-Propyl Alcohol
Unruh JD and Pearson D
Kirk-Othmer Encyclopedia of Chemical Technology (2000)
Evaluation of Taurine by HPTLC reveals the mask of adulterated edible Bird?s nest
Teo P, et al.
Journal of Chemistry, 2013 (2013)
Thermoeconomic and environmental assessment of pressure-swing distillation schemes for the separation of di-n-propyl ether and n-propyl alcohol
Mangili PV
Chemical Engineering and Processing, 148, 107816-107816 (2020)
Extraction of eight triazine and phenylurea herbicides in yogurt by ionic liquid foaming-based solvent floatation
Journal of Chromatography A, 1222, 22-28 (2012)
Effect of short-chain alcohols on surfactant-mediated reversed-phase liquid chromatographic systems
Ruiz-Angel MJ, et al.
Journal of Chromatography A, 1217(45), 7082-7089 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service