Skip to Content
Merck
All Photos(1)

Documents

218832

Sigma-Aldrich

Caspase-3/7 Inhibitor II

The Caspase-3/7 Inhibitor II, also referenced under CAS 775289-20-8, controls the biological activity of Caspase-3/7. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

Caspase-3/7 Inhibitor II, Ac-DNLD-CHO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H31N5O10
CAS Number:
Molecular Weight:
501.49
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

water: 1 mg/mL
DMSO: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A tetrapeptidyl aldehyde that acts as a potent, reversible and active site binding inhibitor of caspases-3 and -7 (IC50 = 3.2 nM and 22.6 nM, respectively) and displays ~100-fold greater selectivity over caspases-8 and -9 (IC50 = 577.6 nM and 364.7 nM, respectively). Offers protection against Camptothecin (Cat. No. 208925), and anti-Fas-mediated apoptosis in Jurkat T cells.
A tetrapeptidyl aldehyde that acts as a potent, reversible and active site binding inhibitor of caspases-3 and -7 (IC50 = 3.2 nM and 22.6 nM, respectively) and displays ~100-fold greater selectivity over caspases-8 and -9 (IC50 = 577.6 nM and 364.7 nM, respectively). Offers protection against Camptothecin (Cat. No. 208925), and anti-Fas-mediated apoptosis in Jurkat T cells.

Note: aldehyde-based inhibitors are less cell-permeable than FMK-based inhibitors, so higher concentrations may be required for cell-based inhibition studies (e.g. 100 µM).

Biochem/physiol Actions

Cell permeable: no
Primary Target
caspase-3, caspase-7
Product does not compete with ATP.
Reversible: yes
Target IC50: 3.2 nM and 22.6 nM, against caspases-3 and -7, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

Ac-Asp-Asn-Leu-Asp-CHO

Analysis Note

Single main spot with additional trace spot by TLC

Other Notes

Yoshimori, A., et al. 2004. BMC Pharmacol.4, 7.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kamila Říhová et al.
Journal of proteome research, 23(8), 2999-3011 (2024-03-19)
Caspase-9 is traditionally considered the initiator caspase of the intrinsic apoptotic pathway. In the past decade, however, other functions beyond initiation/execution of cell death have been described including cell type-dependent regulation of proliferation, differentiation/maturation, mitochondrial, and endosomal/lysosomal homeostasis. As previous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service