Skip to Content
Merck
All Photos(1)

Key Documents

208912

Sigma-Aldrich

Anti-Calreticulin Mouse mAb (FMC75)

liquid, clone FMC75, Calbiochem®

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€446.00

€446.00


Estimated to ship on23 April 2025


Request a Bulk Order

Select a Size

Change View
50 μG
€446.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€446.00


Estimated to ship on23 April 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

FMC75, monoclonal

form

liquid

does not contain

preservative

species reactivity

human, monkey

should not react with

rat, E. coli, canine, hamster, salmon, bovine, mouse, guinea pig

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG1

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Anti-Calreticulin, mouse monoclonal, clone FMC75, recognizes the ~63 kDa calreticulin in Molt 4, MCF-7, HeLa, K562, A431 & other cells. It is validated for FC, WB, ICC, IHC, and in enzyme immunoassay.
Protein G purified mouse monoclonal antibody. Recognizes the ~63 kDa (apparent MW) calreticulin protein.
Recognizes the ~63 kDa (apparent MW) calreticulin protein in Molt 4, MCF-7, HeLa, K562, A431, Jurkat, Hs67, Hs198, and vero cells.

Immunogen

Human
a recombinant protein consisting of human calreticulin fused to maltose binding protein

Application

Enzyme Immunoassay (0.1 µg/ml)

Immunoblotting (0.5 µg/ml, colorimetric; see application references)

Immunocytochemistry (see application references)

Immunohistochemistry (see application references)

Flow Cytometry (see application references)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In PBS, 50% Glycerol and 0.09% Sodium Azide.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
Human Molt 4, MCF-7, HeLa, K562, A431, Jurkat, Hs67, Hs198 cells, or monkey Vero cells

Other Notes

Does not cross-react with GrpE or Grp78. Variables associated with assay conditions will dictate the proper working dilution.
Nauseef, W.M., et al. 1995. J. Biol. Chem. 270, 4741.
Jethmalani, S.M., et al. 1994. J. Biol. Chem. 269, 23603.
Smith, M.J., and Koch, G.L.E. 1989. EMBO J.8, 3581.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephanie S Lam et al.
Nature methods, 12(1), 51-54 (2014-11-25)
APEX is an engineered peroxidase that functions as an electron microscopy tag and a promiscuous labeling enzyme for live-cell proteomics. Because limited sensitivity precludes applications requiring low APEX expression, we used yeast-display evolution to improve its catalytic efficiency. APEX2 is
Ying Yin et al.
PLoS biology, 16(8), e2005140-e2005140 (2018-08-08)
Some secreted proteins that assemble into large complexes, such as extracellular matrices or hormones and enzymes in storage granules, must be kept at subaggregation concentrations during intracellular trafficking. We show surfeit locus protein 4 (Surf4) is the cargo receptor that
Mutation-Driven S100A8 Overexpression Confers Aberrant Phenotypes in Type 1 CALR-Mutated MPN.
Wang, et al.
International Journal of Molecular Sciences, 24 (2023)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service