Skip to Content
MilliporeSigma

192

Crystal Violet

Harleco®, certified by the BSC

Synonym(s):

Gentian Violet, C. I. 42555

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41116121
Colour Index Number:
42555
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


agency

certified by the BSC

Quality Segment

form

solid

manufacturer/tradename

Harleco®

λmax

589-594 nm

application(s)

diagnostic assay manufacturing
hematology
histology

shipped in

ambient

storage temp.

10-30°C

General description

HARLECO® For staining bacteria and spores by Gram′s method

Application

Certified by the Biological Stain Commission (BSC)
For In-Vitro Diagnostic Use.
For Professional Use.
Used for staining bacteria and spores by Gram’s method. Same as Gentian Violet, Catalog #227.
HARLECO® For staining bacteria and spores by Gram′s method

Legal Information

HARLECO is a registered trademark of Merck KGaA, Darmstadt, Germany


signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library