Skip to Content
Merck

Skip To

07-2139-I

Anti-Cysteine Sulfenic Acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
25 μL
Check Cart for Availability
€ 260,00
100 μL
Check Cart for Availability
€ 407,00

About This Item

NACRES:
NA.41
UNSPSC Code:
51111800
Clone:
polyclonal
Species reactivity:
-
Application:
Citations:
-

€ 260,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


clone

polyclonal

Quality Level

isotype

IgG

Immunogen

KLH-conjugated cysteine sulfenic acid.

Biochem/physiol Actions

This rabbit polyclonal antibody detects cysteine sulfenic acid in proteins.

Preparation Note

Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Stable for 1 year at -10°C to -25°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
07-2139-I-25UL04065265237833
07-2139-I-100UL04065265237826