Skip to Content
Merck
All Photos(1)

Key Documents

MAN238929173

Manchester Organics

5-Bromo-2,4-difluorobenzenesulfonamide

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€116.92

Vendor SKU: H14126-25G

€116.92


Ships in 10 days - from Aldrich Partner.


Select a Size

Change View
25 G
€116.92

About This Item

CAS Number:
Molecular Weight:
272.07
MDL number:

Vendor SKU: H14126-25G

€116.92


Ships in 10 days - from Aldrich Partner.

Iupac Name

5-bromo-2,4-difluorobenzenesulfonamide

Inchi Code

1S/C6H4BrF2NO2S/c7-3-1-6(13(10,11)12)5(9)2-4(3)8/h1-2H,(H2,10,11,12)

InChI key

VUBHGNHDBFATNY-UHFFFAOYSA-N

Purity

97%

Country of Origin

GB

Product Link

https://www.manchesterorganics.com/H14126

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service