Skip to Content
Merck
All Photos(1)

Key Documents

MAN182109499

Manchester Organics

3-Acrylamido-3-methylbutyric acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€151.42

Vendor SKU: W18279-1G

€151.42


Ships in 10 days - from Aldrich Partner.


Select a Size

Change View
1 G
€151.42

About This Item

CAS Number:
Molecular Weight:
171.2
MDL number:

Vendor SKU: W18279-1G

€151.42


Ships in 10 days - from Aldrich Partner.

Iupac Name

3-(acryloylamino)-3-methylbutanoic acid

Inchi Code

1S/C8H13NO3/c1-4-6(10)9-8(2,3)5-7(11)12/h4H,1,5H2,2-3H3,(H,9,10)(H,11,12)

InChI key

XOQMWEWYWXJOAN-UHFFFAOYSA-N

Purity

95%

Country of Origin

GB

Product Link

https://www.manchesterorganics.com/W18279

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service