mp
62-65
Iupac Name
2-fluorodecanoic acid
Inchi Code
1S/C10H19FO2/c1-2-3-4-5-6-7-8-9(11)10(12)13/h9H,2-8H2,1H3,(H,12,13)
InChI key
BWUGLYOYLQJRBE-UHFFFAOYSA-N
Purity
95
Country of Origin
UA
Product Link
https://www.enaminestore.com/catalog/EN300-8131703
Melting Point
62-65
MSDS
https://www.enaminestore.com/catalog/EN300-8131703
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service