Skip to Content
Merck
All Photos(1)

Key Documents

ENAH11E138C1

Enamine

tert-butyl 3-(1H-imidazol-4-yl)azetidine-1-carboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
€ 419,40
100 MG
€ 602,70
250 MG
€ 841,90

Vendor SKU: EN300-1894608-50MG

€ 419,40


Availability
Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
50 MG
€ 419,40
100 MG
€ 602,70
250 MG
€ 841,90

About This Item

CAS Number:
Molecular Weight:
223.27
MDL number:

Vendor SKU: EN300-1894608-50MG

€ 419,40


Availability
Ships in 14 days - from Aldrich Partner.

Iupac Name

tert-butyl 3-(1H-imidazol-4-yl)azetidine-1-carboxylate

Inchi Code

1S/C11H17N3O2/c1-11(2,3)16-10(15)14-5-8(6-14)9-4-12-7-13-9/h4,7-8H,5-6H2,1-3H3,(H,12,13)

InChI key

ZFDOKQPYSDPGSA-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-1894608

MSDS

https://www.enaminestore.com/catalog/EN300-1894608


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service