ENA447956503
Enamine
4-(difluoromethoxy)-3-nitropyridine
Select a Size
Vendor SKU: EN300-204831-100MG
€380.40
Select a Size
About This Item
Vendor SKU: EN300-204831-100MG
€380.40
Iupac Name
4-(difluoromethoxy)-3-nitropyridine
Inchi Code
1S/C6H4F2N2O3/c7-6(8)13-5-1-2-9-3-4(5)10(11)12/h1-3,6H
InChI key
FEVJSTPDIPGFNA-UHFFFAOYSA-N
Purity
Country of Origin
Product Link
MSDS
https://www.enaminestore.com/catalog/EN300-204831
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service