ENA432539740
Enamine
2-(2-phenoxypropanamido)acetic acid
Select a Size
Vendor SKU: EN300-749190-100MG
€ 204,90
Select a Size
About This Item
Vendor SKU: EN300-749190-100MG
€ 204,90
Iupac Name
[(2-phenoxypropanoyl)amino]acetic acid
Inchi Code
1S/C11H13NO4/c1-8(11(15)12-7-10(13)14)16-9-5-3-2-4-6-9/h2-6,8H,7H2,1H3,(H,12,15)(H,13,14)
InChI key
HQTYAFXQEWULMT-UHFFFAOYSA-N
storage temp.
Purity
Country of Origin
Shipping Temperature
Physical Form
Product Link
Signal Word
Warning
Hazard Statements
MSDS
https://www.enaminestore.com/catalog/EN300-196752
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service