Skip to Content
Merck
All Photos(1)

Key Documents

ENA390347154

Enamine

2,6-dimethylcyclohex-2-ene-1-carbaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
138.21
MDL number:

Pricing and availability is not currently available.

Iupac Name

2,6-dimethyl-2-cyclohexene-1-carbaldehyde

Inchi Code

1S/C9H14O/c1-7-4-3-5-8(2)9(7)6-10/h4,6,8-9H,3,5H2,1-2H3

InChI key

ZTNFZIHZMITMGE-UHFFFAOYSA-N

storage temp.

4

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

liquid

Product Link

https://www.enaminestore.com/catalog/EN300-125647

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-125647


  • Certificates of Analysis (COA)

    Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

    Need A Sample COA?

    This is a sample Certificate of Analysis (COA) and may not represent a recently manufactured lot of this specific product.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service