Skip to Content
Merck
All Photos(1)

Key Documents

COMH93D5F3C1

Combi-Blocks, Inc.

2-Bromo-3-tetradecylthiophene

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€419.97
5 G
€1,139.91

Vendor SKU: HD-0942-1G

€419.97


Ships in 14 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
1 G
€419.97
5 G
€1,139.91

About This Item

CAS Number:
Molecular Weight:
359.41
MDL number:

Vendor SKU: HD-0942-1G

€419.97


Ships in 14 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.

Iupac Name

2-bromo-3-tetradecylthiophene

Inchi Code

1S/C18H31BrS/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-17-15-16-20-18(17)19/h15-16H,2-14H2,1H3

InChI key

GOUPSYVDGUEWMF-UHFFFAOYSA-N

storage temp.

Refrigerated

Purity

95%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?HD-0942

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?HD-0942


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service