Skip to Content
Merck
All Photos(1)

Key Documents

COM964463885

Combi-Blocks, Inc.

Bicyclo[2.2.2]oct-5-ene-2-carboxaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
136.19
MDL number:

Iupac Name

bicyclo[2.2.2]oct-5-ene-2-carbaldehyde

Inchi Code

1S/C9H12O/c10-6-9-5-7-1-3-8(9)4-2-7/h1,3,6-9H,2,4-5H2

InChI key

KCHFYKPIADOPBJ-UHFFFAOYSA-N

storage temp.

Freezer

Purity

95%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?QV-2445

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?QV-2445


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service