COM466007743
Combi-Blocks, Inc.
(5-Bromothiophen-2-yl)methanamine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
(5-bromo-2-thienyl)methanamine
Inchi Code
1S/C5H6BrNS/c6-5-2-1-4(3-7)8-5/h1-2H,3,7H2
InChI key
RAVKMGSIHFLQJU-UHFFFAOYSA-N
storage temp.
Freezer
Purity
97%
Country of Origin
CN
Product Link
https://www.combi-blocks.com/cgi-bin/find.cgi?QE-8989
MSDS
https://www.combi-blocks.com/cgi-bin/find.cgi?QE-8989
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service