Skip to Content
Merck
All Photos(1)

Key Documents

ATEH75DBF577

AstaTech, Inc.

4-FLUORONICOTINIC ACID

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CA$757.00

CA$757.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
CA$757.00

About This Item

CAS Number:
Molecular Weight:
141.1
MDL number:

CA$757.00


Please contact Customer Service for Availability

Iupac Name

4-fluoronicotinic acid

Inchi Code

1S/C6H4FNO2/c7-5-1-2-8-3-4(5)6(9)10/h1-3H,(H,9,10)

InChI key

UBIVUIKWHBNGFG-UHFFFAOYSA-N

Purity

95%

Physical Form

Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=129020

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=W18401


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service