Skip to Content
Merck
All Photos(1)

Key Documents

ATE517254942

AstaTech, Inc.

METHYL 3-FORMYLCYCLOHEXANE-1-CARBOXYLATE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
170.21
MDL number:

Iupac Name

methyl 3-formylcyclohexane-1-carboxylate

Inchi Code

1S/C9H14O3/c1-12-9(11)8-4-2-3-7(5-8)6-10/h6-8H,2-5H2,1H3

InChI key

OSEJUCOTKIRRNR-UHFFFAOYSA-N

Purity

95%

Physical Form

Colorless to Yellow Liquid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=94769

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=P16243


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service