Skip to Content
Merck
All Photos(1)

Key Documents

APOH03CD8F94

Apollo Scientific Ltd

3-Chloro-4-iodobenzonitrile

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
263.46
MDL number:

Pricing and availability is not currently available.

Iupac Name

3-chloro-4-iodobenzonitrile

Inchi Code

1S/C7H3ClIN/c8-6-3-5(4-10)1-2-7(6)9/h1-3H

InChI key

OUVGLBKTGXKVKS-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/3-chloro-4-iodobenzonitrile

Pictograms

Exclamation mark

Signal Word

Warning

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/3-chloro-4-iodobenzonitrile


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service