Skip to Content
Merck
All Photos(1)

Key Documents

APO456876695

Apollo Scientific Ltd

3-Fluoro-2-methylphenylacetic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€80.70
5 G
€158.80
25 G
€404.10

Vendor SKU: PC0889-1G

€80.70


Ships in 14 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
1 G
€80.70
5 G
€158.80
25 G
€404.10

About This Item

CAS Number:
Molecular Weight:
168.17
MDL number:

Vendor SKU: PC0889-1G

€80.70


Ships in 14 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.

bp

90-92

Iupac Name

(3-fluoro-2-methylphenyl)acetic acid

Inchi Code

1S/C9H9FO2/c1-6-7(5-9(11)12)3-2-4-8(6)10/h2-4H,5H2,1H3,(H,11,12)

InChI key

ZESQUNYKCICBQB-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

98%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid-Crystals

Product Link

https://store.apolloscientific.co.uk/product/3-fluoro-2-methylphenylacetic-acid

Synonym

(3-Fluoro-2-methylphenyl)acetic acid, 2-(3-Fluoro-2-methylphenyl)ethanoic acid, 2-(Carboxymethyl)-6-fluorotoluene

Boiling point

90-92

MSDS

https://store.apolloscientific.co.uk/product/3-fluoro-2-methylphenylacetic-acid


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service