Skip to Content
Merck
All Photos(1)

Documents

APO455837701

Apollo Scientific Ltd

4-(1,1,2,2-Tetrafluoroethoxy)toluene

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
208.16
MDL number:

mp

61.5

density

1.25

Iupac Name

1-methyl-4-(1,1,2,2-tetrafluoroethoxy)benzene

Inchi Code

1S/C9H8F4O/c1-6-2-4-7(5-3-6)14-9(12,13)8(10)11/h2-5,8H,1H3

InChI key

VACIXOPZIXUAKS-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

98%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Liquid

Product Link

https://store.apolloscientific.co.uk/product/4-1122-tetrafluoroethoxytoluene

Flash Point

67-69/14mm

Melting Point

61.5

density

1.25

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/4-1122-tetrafluoroethoxytoluene


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service