Skip to Content
Merck
All Photos(1)

Key Documents

APO455834944

Apollo Scientific Ltd

6-chloro-7-hydroxy-4-(trifluoromethyl)coumarin

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€59.80

Vendor SKU: PC1931C-500MG

€59.80


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
500 MG
€59.80

About This Item

CAS Number:
Molecular Weight:
264.59
MDL number:

Vendor SKU: PC1931C-500MG

€59.80


Ships in 14 days - from Aldrich Partner.

bp

178-182

Iupac Name

6-chloro-7-hydroxy-4-(trifluoromethyl)-2H-chromen-2-one

Inchi Code

1S/C10H4ClF3O3/c11-6-1-4-5(10(12,13)14)2-9(16)17-8(4)3-7(6)15/h1-3,15H

InChI key

AJOANILONFCHFL-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

97%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/6-chloro-7-hydroxy-4-trifluoromethylcoumarin

Synonym

6�hloro-4-trifluoromethylumbelliferone, 6-Chloro-7-hydroxy-4-(trifluoromethyl)-2H-chromen-2-one

Boiling point

178-182

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/6-chloro-7-hydroxy-4-trifluoromethylcoumarin


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service