Skip to Content
Merck
All Photos(1)

Key Documents

APO455833761

Apollo Scientific Ltd

4-Chloro-2-(chloromethyl)-3-methoxypyridine hydrochloride,

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
228.5
MDL number:

Iupac Name

4-chloro-2-(chloromethyl)-3-methoxypyridine hydrochloride

Inchi Code

1S/C7H7Cl2NO.ClH/c1-11-7-5(9)2-3-10-6(7)4-8;/h2-3H,4H2,1H3;1H

InChI key

YWYOLOVNOGZWLY-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

90%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/4-chloro-2-chloromethyl-3-methoxypyridine-hydrochloride

Pictograms

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/4-chloro-2-chloromethyl-3-methoxypyridine-hydrochloride


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service