APO455831129
Apollo Scientific Ltd
2,6-Dichloro-4-nitroanisole
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
bp
97-100
mp
160
Iupac Name
1,3-dichloro-2-methoxy-5-nitrobenzene
Inchi Code
1S/C7H5Cl2NO3/c1-13-7-5(8)2-4(10(11)12)3-6(7)9/h2-3H,1H3
InChI key
GJYVJKPFYCKNEC-UHFFFAOYSA-N
storage temp.
Ambient Temperature
Purity
98%
Country of Origin
GB
Shipping Temperature
Ambient Temperature
Physical Form
Solid
Product Link
https://store.apolloscientific.co.uk/product/26-dichloro-4-nitroanisole
Boiling point
97-100
Melting Point
160
Signal Word
Danger
Hazard Statements
Precautionary statements
MSDS
https://store.apolloscientific.co.uk/product/26-dichloro-4-nitroanisole
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service