Skip to Content
Merck
All Photos(1)

Key Documents

APO455830746

Apollo Scientific Ltd

2-(Chloromethyl)-4,6-dimethoxypyrimidine

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€76.30
1 G
€150.00
5 G
€482.20

Vendor SKU: OR4037-250MG

€76.30


Ships in 14 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
250 MG
€76.30
1 G
€150.00
5 G
€482.20

About This Item

CAS Number:
Molecular Weight:
188.61
MDL number:

Vendor SKU: OR4037-250MG

€76.30


Ships in 14 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.

bp

39-42

Iupac Name

2-(chloromethyl)-4,6-dimethoxypyrimidine

Inchi Code

1S/C7H9ClN2O2/c1-11-6-3-7(12-2)10-5(4-8)9-6/h3H,4H2,1-2H3

InChI key

BWCCPOQIMGGGQX-UHFFFAOYSA-N

storage temp.

Store at 2-8C

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid-Crystals

Product Link

https://store.apolloscientific.co.uk/product/2-chloromethyl-46-dimethoxypyrimidine

Boiling point

39-42

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/2-chloromethyl-46-dimethoxypyrimidine


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service