APO455827705
Apollo Scientific Ltd
Methyl 3,5-diiodo-1H-indole-2-carboxylate
Select a Size
Vendor SKU: OR15451-1G
€338.10
Select a Size
About This Item
Vendor SKU: OR15451-1G
€338.10
bp
209-211
Iupac Name
methyl 3,5-diiodo-1H-indole-2-carboxylate
Inchi Code
1S/C10H7I2NO2/c1-15-10(14)9-8(12)6-4-5(11)2-3-7(6)13-9/h2-4,13H,1H3
InChI key
DSDHHKFKQJKTNJ-UHFFFAOYSA-N
storage temp.
Country of Origin
Shipping Temperature
Physical Form
Product Link
Boiling point
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://store.apolloscientific.co.uk/product/methyl-35-diiodo-1h-indole-2-carboxylate
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service