Skip to Content
Merck
All Photos(1)

Key Documents

AOBH961DC1DF

AOBChem

Methyl 5-bromo-3-ethoxy-2-fluorobenzoate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
277.09
MDL number:

Iupac Name

methyl 5-bromo-3-ethoxy-2-fluorobenzoate

Inchi Code

1S/C10H10BrFO3/c1-3-15-8-5-6(11)4-7(9(8)12)10(13)14-2/h4-5H,3H2,1-2H3

InChI key

UIZCSAOROLZEKA-UHFFFAOYSA-N

Purity

97

Country of Origin

CN

Physical Form

Solid

Product Link

https://aobchem.com/methyl-5-bromo-3-ethoxy-2-fluorobenzoate.html

MSDS

https://aobchem.com/pub/media/files/sds/48056.pdf


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service