AOBH3CD9F472
AOBChem
Bromo-4-(diethoxymethyl)-thiophene,
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
3-bromo-4-(diethoxymethyl)thiophene
Inchi Code
1S/C9H13BrO2S/c1-3-11-9(12-4-2)7-5-13-6-8(7)10/h5-6,9H,3-4H2,1-2H3
InChI key
AKDNDWACGKDMRW-UHFFFAOYSA-N
storage temp.
2-8C
Purity
95
Country of Origin
CN
Shipping Temperature
Room Temperature
Physical Form
Liquid
Product Link
https://aobchem.com/bromo-4-diethoxymethyl-thiophene.html
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://aobchem.com/pub/media/files/sds/32313.pdf
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service