AOBH03B6AB46
AOBChem
2-Fluoro-5-methoxypyridine-4-carboxylic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
2-fluoro-5-methoxyisonicotinic acid
Inchi Code
1S/C7H6FNO3/c1-12-5-3-9-6(8)2-4(5)7(10)11/h2-3H,1H3,(H,10,11)
InChI key
WJWQUZNOYBAAEW-UHFFFAOYSA-N
storage temp.
2-8C
Purity
95
Country of Origin
CN
Shipping Temperature
Room Temperature
Physical Form
Solid
Product Link
https://aobchem.com/2-fluoro-5-methoxypyridine-4-carboxylic-acid.html
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://aobchem.com/pub/media/files/sds/20467.pdf
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service