Skip to Content
Merck
All Photos(1)

Key Documents

AMBH98C8DFA6

Ambeed, Inc.

O-(4-Fluorobenzyl)hydroxylamine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
141.15
MDL number:

Iupac Name

1-[(aminooxy)methyl]-4-fluorobenzene

Inchi Code

1S/C7H8FNO/c8-7-3-1-6(2-4-7)5-10-9/h1-4H,5,9H2

InChI key

PJLRXENMJSXPOU-UHFFFAOYSA-N

storage temp.

Keep in dark place,Inert atmosphere,Store in freezer, under -20C

Purity

95%

Country of Origin

CN

Shipping Temperature

Cold-chain transportation

Physical Form

Solid or liquid

Product Link

https://www.ambeed.com/products/1782-40-7.html

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service