Z680982
Duran® graduated laboratory bottles, without caps
capacity 150 mL, clear glass
Synonym(s):
218012906
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
clear glass
glass cap
narrow-mouth bottle
round bottle
Joint
Joints threaded neck (GL45)
feature
graduations
without cap
packaging
pack of 10 ea
manufacturer/tradename
DURAN® 21 801 29 06
capacity
150 mL
diam. × H
62 mm × 110 mm
Looking for similar products? Visit Product Comparison Guide
General description
DURAN® Original GL laboratory bottles have become the gold standard of multifunctional laboratory bottles since their introduction in 1972. The bottles are moulded from high-purity, clear, USP/EP Type I borosilicate glass 3.3, renowned for its excellent thermal performance, and high resistance to chemical attack. A combination of high-performance materials and robust ISO 4796-1 design provides a long working life. The bottles feature an easy-to-read graduated volume scale, and marking field in permanent white enamel. Permanent Retrace Code on each bottle for manufacturing lot traceability. DURAN™ Original GL bottles are supplied complete with iconic blue polypropylene cap and drip-free pouring ring for use up to 140 °C. All components are fully autoclavable at 121 or 134 °C.
Reusable bottles without caps or pouring rings for the storage of reagents, chemicals, solutions, tissue culture media, etc. Made of Duran borosilicate glass, Type 3.3, for maximum chemical and thermal shock resistance.
Optional pouring rings prevent drips when pouring from these bottles.
Optional pouring rings prevent drips when pouring from these bottles.
Application
Duran™ graduated laboratory bottles, without caps are suitable for the storage of reagents, chemicals, solutions, tissue culture media, and more.
Duran® graduated laboratory bottles, without caps are suitable for the storage of reagents, chemicals, solutions, tissue culture media, and more.
Features and Benefits
- The Duran® graduated laboratory bottles are made of Duran borosilicate glass, Type 3.3, for maximum chemical and thermal shock resistance.
- These bottles are reusable and autoclavable at 121 or 134 °C.
- The optional pouring rings prevent drips when pouring from these bottles.
- The bottles provide an easy-to-read scale with graduation marks to estimate the content.
- They are suitable for long-term storage.
- Complies with the standards in ISO 4796-1:2016 Laboratory glassware -- Bottles -- Part 1: Screw-neck bottles (does not apply to 10 ml bottle)
- Manufactured from DURAN™ borosilicate glass 3.3. DURAN™ is a neutral glass, classified as a Glass Type I according to USP <660>, EP (3.2.1) and JP <7.01>, and meets ASTM E438, Type I, Class A requirements
- Standardised GL-thread and matching cap system for particularly tight sealing and simple, clean pouring out
- Retrace Code, and certificate for manufacturing lot traceability
- Fired-on, highly durable white print
- Suitable for long-term storage
- Easy-to-read scale with graduation marks to estimate the content
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service