Z316520
Stuart™ Scientific Gyro rocker
STR9, speed 10-70 rpm
All Photos(1)
About This Item
Recommended Products
AC/DC input
230 V
grade
CE Compliant
feature
autoclavable
manufacturer/tradename
STR9
availability
not available in USA and Canada
parameter
10-70 rpm speed
platform L × W
360 mm × 360 mm , Fitted with non-slip mat
General description
Unit has a 3-D gyroscopic motion providing a gentle swirling movement which is ideally suited for mixing under low foaming conditions, including DNA extractions, staining and destaining gels, cell culture and general mixing applications.
- Optional 2 deck rack for platelet agitation
- Nylon coated steel, 2 decks
- Adjustable angle of tilt: 0 to 12 degrees.
Legal Information
Stuart is a trademark of Bibby Sterlin Ltd.