Skip to Content
Merck
All Photos(1)

Key Documents

Z183687

Poly-net® protective netting

green, diam. 4-6 in., Close-mesh

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
NACRES:
NB.35

diam.

4-6 in.

thickness

0.07-1.2 mm , approximate

color

green

Looking for similar products? Visit Product Comparison Guide

General description

PE net sleeve stretches around glassware to prevent scratching,chipping or breaking. Netting captures flying glass if container shatters. Easily cut to any length. Color-coded for size identification.

Application

Bottles, columns, and flasks

Legal Information

Poly-net is a registered trademark of Norddeutsche Seekabelwerke GmbH & Co. KG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service