Skip to Content
Merck
All Photos(2)

Key Documents

Z150975

Cochranes Minit size bond tube sets

white, flexible, L 21.0 cm

Synonym(s):

chemistry model sets, molecular model sets

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
€9.20

€9.20


Estimated to ship on26 April 2025



Select a Size

Change View
10 EA
€9.20

About This Item

UNSPSC Code:
60104506
NACRES:
NB.17

€9.20


Estimated to ship on26 April 2025


manufacturer/tradename

Cochranes 0291

L

21.0 cm

color

white, flexible

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service