Key Documents
SYNF311100
Synthware recovery flask
100 mL, joint: ST/NS 14/20
Synonym(s):
Synthware F311100, oval shape flask
Select a Size
1 EA
€24.90
€24.90
Estimated to ship on27 April 2025
Select a Size
Change View
1 EA
€24.90
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
€24.90
Estimated to ship on27 April 2025
material
borosilicate glass flask
pear-shaped
single-neck
Joint
joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F311100
capacity
100 mL
100 mL
General description
Heavy wall design. All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)