SLW3298/02
Pyrex® Burettes, class B, Sofnol, automatic zero, Rotaflo(R) stopcock, PTFE key
10 mL
Synonym(s):
Pyrex, SciLabware 3298/02
All Photos(1)
About This Item
Recommended Products
material
PTFE key
feature
graduations 0.1 mL
manufacturer/tradename
3298/02
SciLabware
capacity
10 mL
General description
- Complies with ISO 385
- With PTFE Rotaflo Stopcock for grease-free performance and fine drop-wise control
- 3mm bore PTFE key
- Graduations in white enamel
- Manufactured from borosilicate glass
- Burettes and bottles should be ordered separately
Easy filling and automatic zero
- Squeeze the polyethylene bottle to fill the burette
- Release pressure to allow excess liquid to drain back into bottle and set the burette to zero
Features and Benefits
• Complies with ISO 385• Manufactured from borosilicate glass 3.3• With PTFE Rotaflo® Stopcock for grease-free performance and fine drop-wise control• 3mm bore PTFE key• Graduations in white enamel• Burettes and bottles should be ordered separately• Easy filling and automatic zero - Squeeze the polyethylene bottle to fill the burette - Release pressure to allow excess liquid to drain back into bottle and set the burette to zero
Legal Information
PYREX is a registered trademark of Corning, Inc.
Rotaflo is a registered trademark of Corning, Inc.