Skip to Content
Merck
All Photos(2)

Documents

DWK261302106

DURAN® test tube

borosilicate glass, tube capacity (18 mL), beaded rim, tube diam. (16 mm) , External Thread

Synonym(s):

261302106

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121700
NACRES:
NB.17

material

borosilicate glass

sterility

sterile; autoclaved

feature

graduations: no

packaging

case of 100 ea

manufacturer/tradename

DURAN® 261302106

tube capacity

(18 mL)

tube diam.

(16 mm) , External Thread

Looking for similar products? Visit Product Comparison Guide

General description

Thick-walled and therefore mechanically very resistant. With round bottom and beaded rim.

Legal Information

DURAN is a registered trademark of DWK Life Sciences

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service