DWK213512308
Duran® Funnel
short stem(s), with print
Synonym(s):
213512308
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
material
short stem(s)
packaging
pkg of 10 ea
manufacturer/tradename
DURAN® 213512308
funnel diam.
35 mm
Looking for similar products? Visit Product Comparison Guide
General description
Transferring liquids and solids is a common working method. In this activity the funnel is used, which due to its shape can absorb a relatively large amount of liquid or solid and transfer it through a constriction into a vessel with a small opening. Funnels are available in different sizes and designs, selected according to their application.
Application
Decanting and filtration of liquids, The spillage of substances is avoided, Filtering, Boro 3.3
Features and Benefits
With short stem; Due to its shape, it is suitable for liquid-solid filtration using a filter paper; Fired-on, highly durable white print
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service