Skip to Content
Merck
All Photos(1)

Documents

CLS954025

Pyrex® Thiele melting point tubes

O.D. × L 25 mm × 150 mm

Synonym(s):

Corning® 9540

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41112203
NACRES:
NB.43

material

borosilicate glass
round bottom tube

packaging

case of 12
pack of 2

manufacturer/tradename

Corning 9540-25

O.D. × L

25 mm × 150 mm

color

clear

General description

For melting point determination tests.

Legal Information

Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service