Skip to Content
Merck
All Photos(4)

Key Documents

H11696

Sigma-Aldrich

Hexamethylenediamine

Hexamethylenediamine

98%

Synonym(s):

1,6-Diaminohexane, 1,6-Hexanediamine, HMDA

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€22.90
100 G
€38.40
500 G
€72.40
1 KG
€131.00

€22.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€22.90
100 G
€38.40
500 G
€72.40
1 KG
€131.00

About This Item

Linear Formula:
NH2(CH2)6NH2
CAS Number:
Molecular Weight:
116.20
Beilstein:
1098307
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
bp:
204-205 °C

€22.90


Check Cart for Availability

Request a Bulk Order

vapor density

4 (vs air)

Quality Level

Assay

98%

form

crystalline solid

expl. lim.

6.3 %

refractive index

n20/D 1.439 (lit.)

pH

12.4 (25 °C, 100 g/L)

bp

204-205 °C

mp

42-45 °C (lit.)

density

0.89 g/mL at 25 °C (lit.)

SMILES string

NCCCCCCN

InChI

1S/C6H16N2/c7-5-3-1-2-4-6-8/h1-8H2

InChI key

NAQMVNRVTILPCV-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Hexamethylenediamine (HMDA) can be used as a reagent used to modify acrylonitrile copolymers[1] and to polycondense with compounds such as calcein.[2]
It can be used as a:
  • Reactant for the synthesis of dialkyl hexamethylene-1,6-dicarbamate with alkyl carbamate in the presence of various metal catalysts via the transesterification reaction.[3]
  • Surface modifier for chlorinated and fluorinated nanodiamond synthesized by detonation.[4]
  • Reactant with adipic acid for the synthesis of nylon 6-6 by polycondensation.[5]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

176.0 °F - closed cup

Flash Point(C)

80 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hexamethylenediamine (HMDA) from fossil-vs. bio-based routes: an economic and life cycle assessment comparative study.
Dros AB, et al.
Green Chemistry, 17(10), 4760-4772 (2015)
Sensors and Actuators B, Chemical, 13, 420-420 (1993)
Surface modification of detonation nanodiamond with ethylenediamine and hexamethylenediamine.
Yakovlev RY, et al.
Mendeleev Communications, 4(22), 213-214 (2012)
Journal of Applied Polymer Science. Applied Polymer Symposium, 54, 355-355 (1994)
Synthesis of dialkyl hexamethylene-1, 6-dicarbamate from 1, 6-hexamethylenediamine and alkyl carbamate over FeCl3 as catalyst.
Guo X, et al.
Catalysis Communications, 10(8), 1248-1251 (2009)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service