Skip to Content
Merck
All Photos(1)

Documents

858951

Sigma-Aldrich

Nε-Methyl-L-lysine hydrochloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H16N2O2 · HCl
CAS Number:
Molecular Weight:
196.68
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

98%

SMILES string

Cl.CNCCCC[C@H](N)C(O)=O

InChI

1S/C7H16N2O2.ClH/c1-9-5-3-2-4-6(8)7(10)11;/h6,9H,2-5,8H2,1H3,(H,10,11);1H/t6-;/m0./s1

InChI key

AQELUQTVJOFFBN-RGMNGODLSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Minami et al.
Journal of biochemistry, 97(3), 745-753 (1985-03-01)
The amino acid sequence of a ferredoxin from a thermoacidophilic archaebacterium, Sulfolobus acidocaldarius, was determined by a combination of various conventional methods to be as follows: Gly-Ile-Asp-Pro-Tyr-Arg-Thr-His-Lys-Pro-Val-Val-Gly-Asp-Ser-Ser-Gly-His- Lys-Ile -Tyr-Gly-Pro-Val-Glu-Ser-Pro-Lys(Me)-Val-Leu-Gly-Val-His-Gly-Thr-Ile-Val -Gly-Va l-Asp-Phe-Asp-Leu-Cys-Ile-Ala-Asp-Gly-Ser-Cys-Ile-Thr-Ala-Cys-Pro-Val-As n-Val-P he-Gln-Trp-Tyr-Glu-Thr-Pro-Gly-His-Pro-Ala-Ser-Glu-Lys-Lys-Ala-Asp-Pro-V al-Asn- Glu-Gln-Ala-Cys-Ile-Phe-Cys-Met-Ala-Cys-Val-Asn-Val-Cys-Pro-Val-Ala-Ala- Ile-Asp -Val-Lys-Pro-Pro. It was composed
A genetically encoded epsilon-N-methyl lysine in mammalian cells.
Dan Groff et al.
Chembiochem : a European journal of chemical biology, 11(8), 1066-1068 (2010-04-28)
K Yoshioka et al.
Journal of bacteriology, 177(4), 1090-1093 (1995-02-01)
We have isolated spontaneous mutants of Salmonella typhimurium which can swim in the presence of antifilament antibodies. The molecular masses of flagellins isolated from these mutants were smaller than that (52 kDa) of wild-type flagellin. Two mutants which produced the
R Garcia et al.
Applied and environmental microbiology, 51(6), 1355-1357 (1986-06-01)
Dansyl derivatives of epsilon-N-mono-, epsilon-N-di-, and epsilon-N-trimethyllysine were resolved from other amino acids in proteins by the use of high-performance liquid chromatography. The system was tested with amino acid standard combinations as well as with acid-hydrolyzed proteins known to contain
F Zappacosta et al.
European journal of biochemistry, 222(3), 761-767 (1994-06-15)
Advanced mass spectrometric procedures have been extensively used to provide an accurate structural characterization of aspartate aminotransferase from Sulfolobus solfataricus. The amino acid sequence of this enzyme had previously been deduced from the DNA sequence. The accurate molecular mass of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service