Skip to Content
Merck
All Photos(1)

Key Documents

763578

Sigma-Aldrich

N,N,N′,N′-Tetraphenylnaphthalene-2,6-diamine

98% (HPLC)

Synonym(s):

NDDP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H26N2
CAS Number:
Molecular Weight:
462.58
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

98% (HPLC)

form

solid

mp

272-276 °C

λmax

312 nm in dichloromethane
341 nm in dichloromethane

SMILES string

c1ccc(cc1)N(c2ccccc2)c3ccc4cc(ccc4c3)N(c5ccccc5)c6ccccc6

InChI

1S/C34H26N2/c1-5-13-29(14-6-1)35(30-15-7-2-8-16-30)33-23-21-28-26-34(24-22-27(28)25-33)36(31-17-9-3-10-18-31)32-19-11-4-12-20-32/h1-26H

InChI key

LEXCBRKPOZULQO-UHFFFAOYSA-N

Application

This material is used as a high performance host material for OLED devices showing increased external quantum efficiency of ~11%.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wu; C.; et al.
Advances in Functional Materials, 19, 3157-3157 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service