Skip to Content
Merck
All Photos(1)

Key Documents

745839

Sigma-Aldrich

ICBA

99.9% (HPLC)

Synonym(s):

1′,1′′,4′,4′′-Tetrahydro-di[1,4]methanonaphthaleno[1,2:2′,3′,56,60:2′′,3′′][5,6]fullerene-C60, C60 derivative, indene-C60 bisadduct

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C78H16
CAS Number:
Molecular Weight:
952.96
UNSPSC Code:
12352103
NACRES:
NA.23

Assay

99.9% (HPLC)

form

solid

mp

236-241 °C

Looking for similar products? Visit Product Comparison Guide

General description

The performance of indene-C60 bisadduct (ICBA) as organic solar cell (OSC) has been described in a study.[1] Simple preparation and high solubility in organic solvents renders ICBA as an efficient substitute to [6,6]-phenyl-C61 butyric acid methyl ester (PCBM).[2]

Application

A ternary bulk heterojunction system (BHJ) may be fabricated with ICBA/ low band gap polymer, for IR sensitized organic solar cells. 1-3ICBA/thiazolo-thiazolo donor polymer blend may be used to enhance the charge generation in BHJ solar cells.[3]
The addition of substituents to a fullerene cage and chemical characterization of the indene-C60 multiadduct gives rise to a reduction of the electron affi nity of the acceptor; resulting in a signifi cant increase in the V oc of devices incorporating these derivatives as the electron acceptor.
Solar Cell (OPV) Device Performance:
ITO/PEDOT:PSS/P3HT:ICBA(1.5:1)/LiF/Al
  • Short-circuit current density (Jsc): 10.53 mA/cm2
  • Open-circuit voltage (Voc): 0.83 V
  • Fill Factor (FF): 0.60
  • Power Conversion Efficiency (PCE): 5.26 %

Legal Information

Plexcore is a registered trademark of Plextronics; Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sih-Hao Liao et al.
Journal of the American Chemical Society, 134(35), 14271-14274 (2012-08-18)
We present a novel electron transport (ET) polymer composed of polyfluorene grafted with a K(+)-intercalated crown ether involving six oxygen atoms (PFCn6:K(+)) for bulk-heterojunction polymer solar cells (PSCs) with regioregular poly(3-hexylthiophene) (P3HT) as the donor and indene-C(60) bisadduct (ICBA) or
Charge Photogeneration for a Series of Thiazolo-Thiazole Donor Polymers Blended with the Fullerene Electron Acceptors PCBM and ICBA.
Shoaee S, et al.
Advances in Functional Materials, 23(26), 3286-3298 (2013)

IR sensitization of an indene-C60 bisadduct (ICBA) in ternary organic solar cells
Ameri T, et al.
Energy & Environmental Science, 6 (2013)
Comparative study of spectral and morphological properties of blends of P3HT with PCBM and ICBA.
Lin YH, et al.
Organic Electronics, 13(11), 2333-2341 (2012)
Beyond PCBM: Understanding the Photovoltaic Performance of Blends of Indene-C60 Multiadducts with Poly(3-hexylthiophene)
Nardes, A. M.; et al.
Advances in Functional Materials, 22, 4115-4127 (2012)

Articles

Fabrication procedure of organic field effect transistor device using a soluble pentacene precursor.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service