Skip to Content
Merck
All Photos(1)

Documents

699985

Sigma-Aldrich

6-Acetyl-1,2,3,4-tetrahydronaphthalene

97%

Synonym(s):

1-(5,6,7,8-Tetrahydronaphthalen-2-yl)ethanone, 6-Acetyltetralin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14O
CAS Number:
Molecular Weight:
174.24
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

liquid

refractive index

n20/D 1.562

density

1.039 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CC(=O)c1ccc2CCCCc2c1

InChI

1S/C12H14O/c1-9(13)11-7-6-10-4-2-3-5-12(10)8-11/h6-8H,2-5H2,1H3

InChI key

VEPUKHYQNXSSKV-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service