Skip to Content
Merck
All Photos(1)

Key Documents

682020

Sigma-Aldrich

2-Benzylimidazo[1,5-a]quinolinium hexafluorophosphate

97%

Synonym(s):

2-(Phenylmethyl)-imidazo[1,5-a]quinolinium hexafluorophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H15N2F6P
CAS Number:
Molecular Weight:
404.29
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

reaction suitability

reagent type: catalyst

mp

148-150 °C

SMILES string

F[P-](F)(F)(F)(F)F.C(c1ccccc1)[n+]2cc3ccc4ccccc4n3c2

InChI

1S/C18H15N2.F6P/c1-2-6-15(7-3-1)12-19-13-17-11-10-16-8-4-5-9-18(16)20(17)14-19;1-7(2,3,4,5)6/h1-11,13-14H,12H2;/q+1;-1

InChI key

VOHJHJRDNQUECO-UHFFFAOYSA-N

Application

Reactant for stable N-heterocyclic carbenes

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service